Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nodal growth differentiation factor | NODAL | HTX5 | Anti-sense | Chr. 10 - 70431936 - 70441709 on NCBI Build GRCh38 | |
| Downstream |
phosphatase domain containing, paladin 1 |
PALD1 |
KIAA1274, PALD |
22.0kb | Sense | Chr. 10 - 70458542 - 70568450 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map