Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | monooxygenase DBH like 1 | MOXD1 |
MOX, PRO5780, dJ248E1.1 |
-54.3kb | Sense | Chr. 6 - 132296055 - 132401534 on NCBI Build GRCh38 |
| Downstream | trace amine associated receptor 9 (gene/pseudogene) | TAAR9 |
TA3, TAR3, TAR9, TRAR3 |
80.3kb | Anti-sense | Chr. 6 - 132538288 - 132539336 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map