Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium voltage-gated channel subfamily H member 7 | KCNH7 |
ERG3, HERG3, Kv11.3 |
-864.3kb | Sense | Chr. 2 - 162371407 - 162838747 on NCBI Build GRCh38 |
| Downstream | growth factor receptor bound protein 14 | GRB14 | 787.6kb | Sense | Chr. 2 - 164492406 - 164621850 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map