Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373805 | LOC105373805 | -121.8kb | Anti-sense | Chr. 2 - 191021526 - 191032314 on NCBI Build GRCh38 | |
| Downstream | myosin IB | MYO1B | myr1 | 89.1kb | Anti-sense | Chr. 2 - 191245381 - 191425389 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map