Assay Details
| Gene Symbol: GPR1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104359.1 | 6 | 3285 | 65 | ||
| GenBank mRNA | AI128443.1 | - | 1 | 65 | ||
| AK002005.1 | - | 1 | 1757 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 1 | GPR1 | -46.0kb | Anti-sense | Chr. 2 - 206175316 - 206218047 on NCBI Build GRCh38 | |
| Downstream | zinc finger DBF-type containing 2 | ZDBF2 | 8.4kb | Sense | Chr. 2 - 206274641 - 206314438 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

GPR1-AS
Genomic Map