Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SERTAD4 antisense RNA 1 | SERTAD4-AS1 | C1orf133 | -10.3kb | Anti-sense | Chr. 1 - 210231459 - 210234121 on NCBI Build GRCh38 |
| Downstream | hedgehog acyltransferase | HHAT |
MART2, SKI1, Skn |
82.0kb | Sense | Chr. 1 - 210327873 - 210676298 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map