Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mir-99a-let-7c cluster host gene | MIR99AHG |
C21orf34, C21orf35, LINC00478, MONC |
-3.8kb | Sense | Chr. 21 - 16070488 - 16631727 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1549 | LINC01549 | C21orf37 | 823.4kb | Sense | Chr. 21 - 17438890 - 17449185 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map