Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PLCG1 antisense RNA 1 | PLCG1-AS1 | TOP1-AS1 | -7.6kb | Anti-sense | Chr. 20 - 41097994 - 41138000 on NCBI Build GRCh38 |
| Downstream | microRNA 6871 | MIR6871 | hsa-mir-6871 | 21.2kb | Sense | Chr. 20 - 41169023 - 41169078 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map