Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth differentiation factor 7 | GDF7 | BMP12 | -72.8kb | Anti-sense | Chr. 2 - 20666664 - 20671490 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928271 | LOC101928271 | 113.9kb | Anti-sense | Chr. 2 - 20859771 - 20861661 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map