Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | F-box and leucine rich repeat protein 13 | FBXL13 |
DRC6, Fbl13 |
Anti-sense | Chr. 7 - 102699226 - 103074841 on NCBI Build GRCh38 | |
| Downstream |
nuclear factor, erythroid 4 |
NFE4 | NF-E4 | 30.1kb | Sense | Chr. 7 - 102973437 - 102978667 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map