Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929441 | LOC101929441 | -232.0 | Anti-sense | Chr. 1 - 204141408 - 204143009 on NCBI Build GRCh38 | |
| Downstream | renin | REN | HNFJ2 | 9.6kb | Sense | Chr. 1 - 204154816 - 204166337 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map