Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADH:ubiquinone oxidoreductase complex assembly factor 7 | NDUFAF7 |
C2orf56, MidA, PRO1853 |
-71.3kb | Anti-sense | Chr. 2 - 37231631 - 37259052 on NCBI Build GRCh38 |
| Downstream | glutaminyl-peptide cyclotransferase | QPCT |
GCT, QC, sQC |
22.6kb | Anti-sense | Chr. 2 - 37344610 - 37373322 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map