Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transient receptor potential cation channel subfamily M member 4 | TRPM4 |
LTrpC4, PFHB1B, TRPM4B, hTRPM4 |
-95.2kb | Anti-sense | Chr. 19 - 49157741 - 49211841 on NCBI Build GRCh38 |
| Downstream | microRNA 4324 | MIR4324 | mir-4324 | 152.0 | Sense | Chr. 19 - 49308797 - 49308868 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map