Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
VPS16, CORVET/HOPS core subunit |
VPS16 | hVPS16 | -109.0kb | Sense | Chr. 20 - 2840697 - 2866732 on NCBI Build GRCh38 |
| Downstream | gonadotropin releasing hormone 2 | GNRH2 |
GnRH-II, LH-RHII |
61.8kb | Sense | Chr. 20 - 3039061 - 3045896 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map