Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | patched 1 pseudogene | LOC100287846 | -483.5kb | Sense | Chr. 8 - 47189307 - 47192816 on NCBI Build GRCh38 | |
| Downstream | CCAAT/enhancer binding protein delta | CEBPD |
C/EBP-delta, CELF, CRP3, NF-IL6-beta |
58.9kb | Anti-sense | Chr. 8 - 47736914 - 47738164 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map