Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CRTC3 antisense RNA 1 | CRTC3-AS1 | -72.9kb | Sense | Chr. 15 - 90620009 - 90717140 on NCBI Build GRCh38 | |
| Downstream |
furin, paired basic amino acid cleaving enzyme |
FURIN |
FUR, PACE, PCSK3, SPC1 |
77.9kb | Anti-sense | Chr. 15 - 90868592 - 90883458 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map