Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elastin microfibril interfacer 2 | EMILIN2 |
EMILIN-2, FOAP-10 |
Sense | Chr. 18 - 2847030 - 2914092 on NCBI Build GRCh38 | |
| Downstream | lipin 2 | LPIN2 | 1.0kb | Anti-sense | Chr. 18 - 2916994 - 3013315 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map