Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAH-box helicase 29 | DHX29 | DDX29 | -113.8kb | Sense | Chr. 5 - 55256245 - 55307693 on NCBI Build GRCh38 |
| Downstream | phospholipid phosphatase 1 | PLPP1 |
LLP1a, LPP1, PAP-2a, PAP2, PPAP2A |
1.0kb | Sense | Chr. 5 - 55424842 - 55535370 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map