Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1139 | LINC01139 | -761.8kb | Sense | Chr. 1 - 238480384 - 238486017 on NCBI Build GRCh38 | |
| Downstream | cholinergic receptor muscarinic 3 | CHRM3 |
EGBRS, HM3, PBS |
135.7kb | Anti-sense | Chr. 1 - 239386565 - 239915450 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map