Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | URB1 antisense RNA 1 (head to head) | URB1-AS1 |
C21orf119, PRED84 |
-47.8kb | Sense | Chr. 21 - 32393133 - 32393960 on NCBI Build GRCh38 |
| Downstream | t-complex 10-like | TCP10L |
C21orf77, PRED77, TCP10A-2 |
132.2kb | Anti-sense | Chr. 21 - 32574841 - 32585535 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map