Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8066 | MIR8066 | hsa-mir-8066 | -56.1kb | Sense | Chr. 4 - 101240795 - 101240872 on NCBI Build GRCh38 |
| Downstream | microRNA 1255a | MIR1255A |
MIRN1255A, hsa-mir-1255a |
32.4kb | Sense | Chr. 4 - 101330302 - 101330414 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map