Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DAB2, clathrin adaptor protein |
DAB2 |
DOC-2, DOC2 |
Anti-sense | Chr. 5 - 39371674 - 39425233 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101926940 | LOC101926940 | 130.1kb | Sense | Chr. 5 - 39520431 - 39524708 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map