Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZNF337 antisense RNA 1 | ZNF337-AS1 | Anti-sense | Chr. 20 - 25624045 - 25678074 on NCBI Build GRCh38 | ||
| Downstream | zinc finger protein 337 | ZNF337 | 13.8kb | Sense | Chr. 20 - 25673195 - 25696904 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map