Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MRS2 pseudogene 2 | MRS2P2 | -75.8kb | Sense | Chr. 12 - 71848292 - 71850983 on NCBI Build GRCh38 | |
| Downstream | tryptophan hydroxylase 2 | TPH2 |
ADHD7, NTPH |
11.6kb | Anti-sense | Chr. 12 - 71938846 - 72032441 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map