Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cysteine and tyrosine rich 1 antisense RNA 1 | CYYR1-AS1 | Anti-sense | Chr. 21 - 26393635 - 26569252 on NCBI Build GRCh38 | ||
| Downstream | ADAM metallopeptidase with thrombospondin type 1 motif 1 | ADAMTS1 |
C3-C5, METH1 |
364.6kb | Sense | Chr. 21 - 26836287 - 26845409 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map