Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ecotropic viral integration site 5 | EVI5 |
EVI-5, NB4S |
Anti-sense | Chr. 1 - 92508696 - 92792404 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein L5 | RPL5 |
DBA6, L5, MSTP030, PPP1R135 |
157.8kb | Sense | Chr. 1 - 92832037 - 92841924 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map