Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apoptosis and caspase activation inhibitor | AVEN | PDCD12 | Sense | Chr. 15 - 33858602 - 34074877 on NCBI Build GRCh38 | |
| Downstream | cholinergic receptor muscarinic 5 | CHRM5 | HM5 | 98.4kb | Anti-sense | Chr. 15 - 33968314 - 34065094 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map