Assay Details
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | L37711.1 | - | 1 | 49 | Chr. 20 - 1379189 - 1379351 [-] on Build GRCh38 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FKBP1A-SDCBP2 readthrough (NMD candidate) | FKBP1A-SDCBP2 | Sense | Chr. 20 - 1309909 - 1393172 on NCBI Build GRCh38 | ||
| Downstream | microRNA 6869 | MIR6869 | hsa-mir-6869 | 13.5kb | Sense | Chr. 20 - 1392900 - 1392961 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map