Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 134 member C | FAM134C | Anti-sense | Chr. 17 - 42579508 - 42609427 on NCBI Build GRCh38 | ||
| Downstream | tubulin gamma 1 | TUBG1 |
CDCBM4, GCP-1, TUBG, TUBGCP1 |
10.2kb | Sense | Chr. 17 - 42609340 - 42615238 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map