| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000358064.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000537853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000539745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000540565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
ENST00000541592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010011.hg.1 |
NM_175054.2 |
Refseq |