Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC285692 | LOC285692 | -267.2kb | Sense | Chr. 5 - 9641315 - 9903824 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 173 member B | FAM173B | JS-2 | 54.3kb | Sense | Chr. 5 - 10225507 - 10249909 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map