Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724579 | LOC102724579 | -88.7kb | Sense | Chr. 2 - 85213652 - 85218169 on NCBI Build GRCh38 | |
| Downstream | trans-golgi network protein 2 | TGOLN2 |
TGN38, TGN46, TGN48, TGN51, TTGN2 |
10.2kb | Anti-sense | Chr. 2 - 85318018 - 85328296 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map