Assay Details
| Gene Symbol: PUF60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136033.2 | NP_001129505.1 | 4 - 5 | 87 | ||
| NM_001271098.1 | NP_001258027.1 | 4 - 5 | 87 | |||
| NM_001271099.1 | NP_001258028.1 | 3 - 4 | 87 | |||
| NM_078480.2 | NP_510965.1 | 4 - 5 | 87 | |||
| XM_011516929.2 | XP_011515231.1 | 5 - 6 | 87 | |||
| XM_017013234.1 | XP_016868723.1 | 4 - 5 | 87 | |||
| XM_017013235.1 | XP_016868724.1 | 4 - 5 | 87 | |||
| XM_017013237.1 | XP_016868726.1 | 5 - 6 | 87 | |||
| XM_017013238.1 | XP_016868727.1 | 5 - 6 | 87 | |||
| XM_017013239.1 | XP_016868728.1 | 4 - 5 | 87 | |||
| GenBank mRNA | AF114818.1 | - | 4 - 5 | 87 | ||
| AF190744.1 | - | 4 - 5 | 87 | |||
| AK055941.1 | - | 3 - 4 | 87 | |||
| AK292373.1 | - | 4 - 5 | 87 | |||
| AL522753.3 | - | 4 - 5 | 87 | |||
| BC011265.1 | - | 4 - 5 | 4155549 | 87 | ||
| BC011979.1 | - | 2 - 3 | 3863660 | 87 | ||
| BC035435.2 | - | 3 - 4 | 87 | |||
| CN371576.1 | - | 3 - 4 | 87 | |||
| CR457143.1 | - | 4 - 5 | 87 | |||
| CT000277.1 | - | 3 - 4 | 87 | |||
More Information
Set Membership: |
|

PUF60
Genomic Map