Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LDL receptor related protein 12 | LRP12 | ST7 | -1.1mb | Sense | Chr. 8 - 104489231 - 104589024 on NCBI Build GRCh38 |
| Downstream | ZFPM2 antisense RNA 1 | ZFPM2-AS1 | 90.5kb | Sense | Chr. 8 - 105780410 - 106060503 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map