Assay Details
| Gene Symbol: LIFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009462.1 | XP_016864951.1 | 1 | 88 | 65 | |
| GenBank mRNA | U78628.1 | - | 1 | 154 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LIFR antisense RNA 1 | LIFR-AS1 | Anti-sense | Chr. 5 - 38556786 - 38671216 on NCBI Build GRCh38 | ||
| Downstream | OSMR antisense RNA 1 (head to head) | OSMR-AS1 | CTD-2127H9.1 | 98.2kb | Sense | Chr. 5 - 38693213 - 38845829 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009462.1 transcripts.
Set Membership: |
|

LIFR
Genomic Map