Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507464 | LOC100507464 | -503.7kb | Anti-sense | Chr. 8 - 49496763 - 49512180 on NCBI Build GRCh38 | |
| Downstream | peroxidasin like | PXDNL |
PMR1, PRM1, VPO2 |
1.3mb | Anti-sense | Chr. 8 - 51319575 - 51811031 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map