Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DIM1 dimethyladenosine transferase 1 homolog | DIMT1 |
DIM1, DIMT1L, HSA9761, HUSSY5 |
Sense | Chr. 5 - 62388523 - 62403939 on NCBI Build GRCh38 | |
| Downstream | importin 11 | IPO11 | RanBP11 | 14.4kb | Anti-sense | Chr. 5 - 62412746 - 62628589 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map