Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protease, serine 2 |
PRSS2 |
TRY2, TRY8, TRYP2 |
-16.0kb | Sense | Chr. 7 - 142770943 - 142774564 on NCBI Build GRCh38 |
| Downstream | T cell receptor beta constant 1 | TRBC1 |
BV05S1J2.2, TCRB, TCRBC1 |
574.0 | Sense | Chr. 7 - 142791694 - 142793141 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map