Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MORN repeat containing 5 | MORN5 |
C9orf113, C9orf18 |
Sense | Chr. 9 - 122159661 - 122200088 on NCBI Build GRCh38 | |
| Downstream | LIM homeobox 6 | LHX6 | LHX6.1 | 21.9kb | Anti-sense | Chr. 9 - 122185626 - 122228893 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map