Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TCR gamma alternate reading frame protein | TARP |
CD3G, TCRG, TCRGC1, TCRGC2, TCRGV |
-10.9kb | Sense | Chr. 7 - 38239577 - 38276741 on NCBI Build GRCh38 |
| Downstream | T cell receptor gamma locus antisense RNA 1 | TRG-AS1 | TCRGV | 24.4kb | Anti-sense | Chr. 7 - 38341577 - 38378637 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map