Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 520 | LINC00520 | C14orf34 | -474.0kb | Sense | Chr. 14 - 55781135 - 55796674 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927690 | LOC101927690 | 243.0kb | Anti-sense | Chr. 14 - 56514331 - 56551309 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map