Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ARRDC3 antisense RNA 1 | ARRDC3-AS1 | -2.0mb | Anti-sense | Chr. 5 - 91380347 - 91420715 on NCBI Build GRCh38 | |
| Downstream | nuclear receptor subfamily 2 group F member 1 | NR2F1 |
BBOAS, BBSOAS, COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1 |
154.7kb | Anti-sense | Chr. 5 - 93583337 - 93594613 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map