Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 14 open reading frame 159 | C14orf159 | -32.9kb | Sense | Chr. 14 - 91114013 - 91225396 on NCBI Build GRCh38 | |
| Downstream | coiled-coil domain containing 88C | CCDC88C |
DAPLE, HKRP2, KIAA1509, SCA40 |
12.3kb | Anti-sense | Chr. 14 - 91271323 - 91418498 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map