Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 641 | ZNF641 | Sense | Chr. 12 - 48334380 - 48351414 on NCBI Build GRCh38 | ||
| Downstream | chromosome 12 open reading frame 54 | C12orf54 |
HSD-29, HSD-30 |
109.7kb | Anti-sense | Chr. 12 - 48413154 - 48496514 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map