Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoglucomutase 1 | PGM1 |
CDG1T, GSD14 |
-251.5kb | Sense | Chr. 1 - 63593276 - 63660245 on NCBI Build GRCh38 |
| Downstream | ROR1 antisense RNA 1 | ROR1-AS1 | 182.3kb | Anti-sense | Chr. 1 - 64094442 - 64113791 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map