Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
serine peptidase inhibitor, Kunitz type 1 |
SPINT1 |
HAI, HAI1, MANSC2 |
-237.0 | Sense | Chr. 15 - 40844018 - 40857655 on NCBI Build GRCh38 |
| Downstream | ras homolog family member V | RHOV |
ARHV, CHP, WRCH2 |
14.0kb | Anti-sense | Chr. 15 - 40872214 - 40874289 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map