Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PROSER2 antisense RNA 1 | PROSER2-AS1 | -124.3kb | Sense | Chr. 10 - 11849608 - 11894710 on NCBI Build GRCh38 | |
| Downstream | dehydrogenase E1 and transketolase domain containing 1 | DHTKD1 |
AMOXAD, CMT2Q |
49.4kb | Anti-sense | Chr. 10 - 12068917 - 12123228 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map