Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373991 | LOC105373991 | -5.2kb | Sense | Chr. 2 - 236553699 - 236569257 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC93463 | LOC93463 | 473.4kb | Anti-sense | Chr. 2 - 237048599 - 237056167 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map