Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105375240 | LOC105375240 | -16.5kb | Anti-sense | Chr. 7 - 39551638 - 39566284 on NCBI Build GRCh38 | |
| Downstream | RALA Ras like proto-oncogene A | RALA | RAL | 39.6kb | Sense | Chr. 7 - 39622955 - 39708124 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map