Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) binding protein cytoplasmic 1 | PABPC1 |
PAB1, PABP, PABP1, PABPC2, PABPL1 |
-190.2kb | Sense | Chr. 8 - 100702916 - 100722998 on NCBI Build GRCh38 |
| Downstream | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta | YWHAZ |
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD |
4.8kb | Sense | Chr. 8 - 100918576 - 100954068 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map